Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mascon Medical PD-Phi29 DNA polymerase, 5000U/pk
SDP

Catalog No. NC3983169
Encompass_Preferred

PD-Phi29 is an engineered variant of the phi29 DNA polymerase with enhanced thermostability and activity compared to wild-type enzyme. The polymerase retains its intrinsic 3´→5' exonuclease activity, allowing high-fidelity proofreading during nucleotide incorporation. PD-phi29 is ideal for isothermal DNA amplification with exceptionally low template input (~1fg DNA). Compatible formats include RCA and MDA for whole genome amplification and plasmid sequencing.

Features:

• Thermostable up to 42°C
• High yield even from ultra-low input
• Low error rate
• Robust strand-displacement processivity
Includes two tubes (enzyme + buffer); store at –15°C to –25°C. For Research Use Only.
PD-Phi29 DNA Polymerase, 0.1 mg/mL, 10,000 U/mL, 0.5 mL (5,000 U/PK)

Catalog No. NC3983169 Supplier Mascon Medical Supplier No. PD00002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PD-Phi29 is an engineered variant of the phi29 DNA polymerase with enhanced thermostability and activity compared to wild-type enzyme. The polymerase retains its intrinsic 3´→5' exonuclease activity, allowing high-fidelity proofreading during nucleotide incorporation. PD-phi29 is ideal for isothermal DNA amplification with exceptionally low template input (~1fg DNA). Compatible formats include RCA and MDA for whole genome amplification and plasmid sequencing.

Features:

• Thermostable up to 42°C
• High yield even from ultra-low input
• Low error rate
• Robust strand-displacement processivity
Includes two tubes (enzyme + buffer); store at –15°C to –25°C. For Research Use Only.
PD-Phi29 DNA Polymerase, 0.1 mg/mL, 10,000 U/mL, 0.5 mL (5,000 U/PK)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.