Learn More
Chemglass Life Sciences Set PD-Tips II, 20pcs of Each Size: 0.5, 1, 1.25, 2.5, 5, 10 and 12.5ml, Non-Sterile, Bulk, Type Encoded
SDP
Catalog No. 501979469
Encompass_Preferred
Set PD-Tips II, 20pcs of Each Size: 0.5, 1, 1.25, 2.5, 5, 10 and 12.5ml, Non-Sterile, Bulk, Type Encoded. BRAND PD-Tip™ II Precision Dispenser Tips offer efficient, step-by-step dispensing and enhance the proven features of the classic BRAND PD-Tip™ syringe tips. Patented size coding means automatic tip volume detection by the HandyStep touch and HandyStep electronic. They also fit repeating pipettes from other manufacturers that use standard sized syringe tips. PD-Tip™ II tips are manufactured from virgin polypropylene and high-density polyethylene. Available in non-sterile and BIO-CERT sterile. BIO-CERT PD-Tip™ II Precision Dispenser Tips are sterile and certified free of DNA, RNase, endotoxins, and ATP to meet the most stringent sample integrity requirements.
Catalog No. 50-197-9469 Supplier Chemglass Life Sciences Supplier No. CLS3468N050
Set PD-Tips II, 20pcs of Each Size: 0.5, 1, 1.25, 2.5, 5, 10 and 12.5ml, Non-Sterile, Bulk, Type Encoded. BRAND PD-Tip™ II Precision Dispenser Tips offer efficient, step-by-step dispensing and enhance the proven features of the classic BRAND PD-Tip™ syringe tips. Patented size coding means automatic tip volume detection by the HandyStep touch and HandyStep electronic. They also fit repeating pipettes from other manufacturers that use standard sized syringe tips. PD-Tip™ II tips are manufactured from virgin polypropylene and high-density polyethylene. Available in non-sterile and BIO-CERT sterile. BIO-CERT PD-Tip™ II Precision Dispenser Tips are sterile and certified free of DNA, RNase, endotoxins, and ATP to meet the most stringent sample integrity requirements.