Learn More
Medchemexpress LLC HY-10254 100mg Medchemexpress, PD0325901 CAS:391210-10-9 Purity:>98%
SDP
Catalog No. 502028712
Encompass_Preferred
Medchemexpress, HY-10254 100mg PD0325901 CAS:391210-10-9 PD0325901 (PD325901) is an orally active, selective and non-ATP-competitive MEK inhibitor with an IC50 of 0.33 nM. PD0325901 exhibits a Kiapp of 1 nM against activated MEK1 and MEK2. PD0325901 suppresses the expression of p-ERK1/2 and induces apoptosis. PD0325901 has anti-cancer activity for a broad spectrum of human tumor xenografts[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8712 Supplier Medchemexpress LLC Supplier No. HY10254100MG
Medchemexpress, HY-10254 100mg PD0325901 CAS:391210-10-9 PD0325901 (PD325901) is an orally active, selective and non-ATP-competitive MEK inhibitor with an IC50 of 0.33 nM. PD0325901 exhibits a Kiapp of 1 nM against activated MEK1 and MEK2. PD0325901 suppresses the expression of p-ERK1/2 and induces apoptosis. PD0325901 has anti-cancer activity for a broad spectrum of human tumor xenografts[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.