Learn More
Enzo Life Sciences PD0325901, (10mg), CAS Number: 391210-10-9
SDP
Catalog No. NC1362612
Encompass_Preferred
Selective MEK inhibitor. PD0325901 (PD325901) is selective and non ATP-competitive MEK inhibitor with an IC50 of 0.33 nM, roughly 500-fold more potent than CI-1040 on the phosphorylation of ERK1 and ERK2. Alternative Name: (R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamide. Formula: C16H14F3IN2O4. MW: 482.2. CAS Number: 391210-10-9. Purity: ≥99% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (96mg/ml) or ethanol (40mg/ml). Long Term Storage: -20°C
Catalog No. NC1362612 Supplier Enzo Life Sciences Supplier No. ENZCHM2020010
Selective MEK inhibitor. PD0325901 (PD325901) is selective and non ATP-competitive MEK inhibitor with an IC50 of 0.33 nM, roughly 500-fold more potent than CI-1040 on the phosphorylation of ERK1 and ERK2. Alternative Name: (R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamide. Formula: C16H14F3IN2O4. MW: 482.2. CAS Number: 391210-10-9. Purity: ≥99% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (96mg/ml) or ethanol (40mg/ml). Long Term Storage: -20°C