Learn More
STEMCELL Technologies PD0325901, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72184
Encompass
MEK/ERK pathway inhibitor; Inhibits MEK
Catalog No. NC1267211
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov