Learn More
Apexbio Technology LLC PD0325901 391210-10-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064107
Encompass_Preferred
PD0325901 (CAS 391210-10-9) is a small-molecule inhibitor selectively targeting mitogen-activated protein kinase kinase (MEK) an integral kinase within the RAS/RAF/MEK/ERK signaling cascade This pathway commonly exhibits elevated activity in various human cancers and contributes to cell proliferation survival and differentiation Treatment with PD0325901 reduces phosphorylated ERK (P-ERK) levels in vitro indicative of MEK inhibition In mouse tumor models administration of PD0325901 has demonstrated suppression of tumor growth underscoring its value as a research tool for elucidating MEK-mediated signaling and evaluating therapeutic potential in oncology
Catalog No. 50-000-64107 Supplier Apexbio Technology LLC Supplier No. A30135.1
PD0325901 (CAS 391210-10-9) is a small-molecule inhibitor selectively targeting mitogen-activated protein kinase kinase (MEK) an integral kinase within the RAS/RAF/MEK/ERK signaling cascade This pathway commonly exhibits elevated activity in various human cancers and contributes to cell proliferation survival and differentiation Treatment with PD0325901 reduces phosphorylated ERK (P-ERK) levels in vitro indicative of MEK inhibition In mouse tumor models administration of PD0325901 has demonstrated suppression of tumor growth underscoring its value as a research tool for elucidating MEK-mediated signaling and evaluating therapeutic potential in oncology