Learn More
Apexbio Technology LLC MEK inhibitor 391210-10-9. MFCD08435926
SDP
Supplier:  Apexbio Technology LLC A301325
Encompass_Preferred
PD0325901 is a specific inhibitor of mitogen-activated protein kinase MEK. PD0325901 is a small molecular with the formula of C16H14F3IN2O4 and Molecular Weight of 482. MEK is a key component of the RAS/RAF/MEK/ERK signaling pathway that is frequently activated in human tumors, and MEK/ERK regulates cell proliferation, survival, and differentiation in response to extracellular signals. PD0325901 effectively reduces P-ERK levels and cell growth in vitro, and inhibits tumor growth in mouse model in vivo. Other sizes are available. Please inqury us for quote.
Catalog No. 50-101-3147