Learn More
Adooq Bioscience LLC PD0325901, 5MG
Supplier:  Adooq Bioscience LLC A102565
Encompass_Preferred
PD0325901 is MEK inhibitor and non-competitive with ATP, Kiapp of 1 nM against activated MEK1 and MEK2.
Catalog No. NC0808832