Learn More
Selleck Chemical LLC Mirdametinib (PD0325901) 5mg 391210-10-9
SDP
Supplier:  Selleck Chemical LLC S10365MG
Encompass_Preferred
Mirdametinib (PD0325901) is a selective and non ATP-competitive MEK inhibitor with IC50 of 0.33 nM in cell-free assays, roughly 500-fold more potent than CI-1040 on phosphorylation of ERK1 and ERK2. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-850-6