Learn More
Abcam PD0332991, Cdk4/Cdk6 inhibitor, 5MG
Catalog No. 502657504
Encompass_Preferred
MW 447.5 Da, Purity >98%. Potent, highly specific inhibitor of Cdk4 (IC₅₀ = 11 nM) and Cdk6 (IC₅₀ = 16 nM). Displays selectivity for Cdk4/6 over a panel of 36 additional protein kinases (IC₅₀ values are > 10 μM). Induces G₁ cell cycle arrest and senescence in retinoblastoma protein (Rb)-proficient cell lines. Blocks growth of intracranial glioblastoma multiforme xenografts in mice. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7504 Supplier Abcam Supplier No. AB2181185MG
MW 447.5 Da, Purity >98%. Potent, highly specific inhibitor of Cdk4 (IC₅₀ = 11 nM) and Cdk6 (IC₅₀ = 16 nM). Displays selectivity for Cdk4/6 over a panel of 36 additional protein kinases (IC₅₀ values are > 10 μM). Induces G₁ cell cycle arrest and senescence in retinoblastoma protein (Rb)-proficient cell lines. Blocks growth of intracranial glioblastoma multiforme xenografts in mice. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement