Learn More
CAPRICO BIOTECHNOLOGIES INC PD1 APC/IFluor700
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4252175
Encompass_Preferred
The PD-1 (APC) iFluor 700 antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes human PD-1. It is conjugated to APC-iFluor 700 and is suitable for flow cytometry, immunohistochemistry, and ELISA. This antibody has been tested and validated for flow cytometry using human peripheral blood cells. Each vial contains 100 tests of antibody in PBS with 0.1% sodium azide and 0.2% BSA. This antibody should be stored at 4°C protected from light.
Catalog No. 50-227-9310