Learn More
PD1 Detection Set, ProSci

Description
PD1 Monoclonal specifically detects PD1 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | PD1 |
| Applications | ELISA |
| Concentration | Antibody 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PD-1 Antibodies are supplied in PBS. |
| Host Species | Llama |
| Immunogen | PD-1 antibodies were raised against a recombinant protein corresponding to amino acids 25 - 167 of human PD-1. |
| Purification Method | PD-1 Antibodies are affinity chromatography purified via Nickel column. Antibodies are supplied as a His-tagged purified protein. They also contains a myc-tag for detection. |
| Quantity | 1 Set |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.