Learn More
Sino Biological Recombinant Anti-PD-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 70109R039100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine PD1 / PDCD1 / CD279 ( Catalog#70109-D08H; XP_022265285.1; Met1-Leu169).
Catalog No. 50-224-8031