Learn More
Apexbio Technology LLC PD123319 130663-39-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22065.1
Encompass_Preferred
PD123319 (CAS 130663-39-7) is a non-peptide antagonist targeting angiotensin II (Ang II) receptors particularly the AT2 receptor subtype It binds selectively exhibiting an IC50 of 34 nM in rat adrenal assays and 210 nM in rat brain binding assays Additionally PD123319 inhibits Ang II binding to bovine adrenal glomerulosa microsomal preparations with an IC50 of 6 9 nM Its antagonistic activity leads to reduced cyclic GMP synthesis and elevated prostaglandin E2 production without altering protein tyrosine phosphorylation or thymidine incorporation mediated by Ang II PD123319 serves as a useful pharmacological tool in investigating Ang II receptor signaling pathways
Catalog No. 50-000-62607