Learn More
TARGETMOL CHEMICALS INC PD128907 HCl 5MG
SDP
Catalog No. 502879940
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. PD128907 HCl is a potent and selective dopamine D3 receptor agonist, with EC50 of 0.64 nM, exhibits 53-fold selectivity over dopamine D2 receptor.
Catalog No. 50-287-9940 Supplier TARGETMOL CHEMICALS INC Supplier No. T61425MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. PD128907 HCl is a potent and selective dopamine D3 receptor agonist, with EC50 of 0.64 nM, exhibits 53-fold selectivity over dopamine D2 receptor.