Learn More
TARGETMOL CHEMICALS INC PD146176 25MG
SDP
Catalog No. 502579183
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD146176 (NSC-168807) is an inhibitor of 15-Lipoxygenase (15-LO) it inhibits rabbit reticulocyte 15-LO with Ki of 197 nM and IC50 of 0.54 uM. PD146176 reverses cognitive impairment brain amyloidosis and tau pathology by stimulating autophagy in aged triple transgenic mice. purity: 99%
Catalog No. 50-257-9183 Supplier TARGETMOL CHEMICALS INC Supplier No. T753025MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD146176 (NSC-168807) is an inhibitor of 15-Lipoxygenase (15-LO) it inhibits rabbit reticulocyte 15-LO with Ki of 197 nM and IC50 of 0.54 uM. PD146176 reverses cognitive impairment brain amyloidosis and tau pathology by stimulating autophagy in aged triple transgenic mice. purity: 99%