Learn More
Medchemexpress LLC HY-100529 10mg , PD150606 CAS:179528-45-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10052910MG
Encompass_Preferred
HY-100529 10mg PD150606 CAS: 179528-45-1 PD 150606 is a selective, cell-permeable non-peptide calpain inhibitor with Ki values of 0.21 μM and 0.37 μM for μ- and m-calpains respectively, which is neuroprotective. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0373