Learn More
Medchemexpress LLC HY-100529 5mg Medchemexpress, PD150606 CAS:179528-45-1 Purity:>98%
SDP
Catalog No. 501931870
Encompass_Preferred
Medchemexpress, HY-100529 5mg PD150606 CAS:179528-45-1 PD 150606 is a selective, cell-permeable non-peptide calpain inhibitor with K i values of 0.21 μM and 0.37 μM for μ- and m-calpains respectively, which is neuroprotective [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1870 Supplier Medchemexpress LLC Supplier No. HY1005295MG
Medchemexpress, HY-100529 5mg PD150606 CAS:179528-45-1 PD 150606 is a selective, cell-permeable non-peptide calpain inhibitor with K i values of 0.21 μM and 0.37 μM for μ- and m-calpains respectively, which is neuroprotective [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.