Learn More
Selleck Chemical LLC PD153035 HCl 10mg 183322-45-4 SU-5271 (AG1517) HCl ,ZM 252868 HCl
SDP
Catalog No. 505869
Encompass_Preferred
PD153035 HCl (SU-5271 HCl, AG1517 HCl, ZM 252868 HCl) is a potent and specific inhibitor of EGFR with Ki and IC50 of 5.2 pM and 29 pM in cell-free assays; little effect noted against PGDFR, FGFR, CSF-1, InsR and Src. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-586-9 Supplier Selleck Chemical LLC Supplier No. S107910MG
PD153035 HCl (SU-5271 HCl, AG1517 HCl, ZM 252868 HCl) is a potent and specific inhibitor of EGFR with Ki and IC50 of 5.2 pM and 29 pM in cell-free assays; little effect noted against PGDFR, FGFR, CSF-1, InsR and Src. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.