Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PD153035 hydrochloride | 183322-45-4 | 99.5% | 1 ML
SDP

Catalog No. 5000233940
Encompass_Preferred

PD153035 hydrochloride is a potent ATP-competitive epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor supplied as a ready-to-use DMSO stock solution for biochemical and cell-based research applications.

  • Potent EGFR inhibitor with reported Ki = 6 pM and IC50 = 25 pM.
  • Supplied as a 10 mM solution in DMSO (1 mL) for convenient dosing in assays.
  • High purity suitable for biochemical and cell-based studies.
  • CAS number 183322-45-4 for unambiguous identification.
  • Molecular weight 396.67 g/mol and formula C16H15BrClN3O2.
  • Widely used in kinase signaling and cancer research to block EGFR activity.

Catalog No. 50-002-33940 Supplier Medchemexpress LLC Supplier No. HY1201310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PD153035 hydrochloride is a potent ATP-competitive epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor supplied as a ready-to-use DMSO stock solution for biochemical and cell-based research applications.

  • Potent EGFR inhibitor with reported Ki = 6 pM and IC50 = 25 pM.
  • Supplied as a 10 mM solution in DMSO (1 mL) for convenient dosing in assays.
  • High purity suitable for biochemical and cell-based studies.
  • CAS number 183322-45-4 for unambiguous identification.
  • Molecular weight 396.67 g/mol and formula C16H15BrClN3O2.
  • Widely used in kinase signaling and cancer research to block EGFR activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.