Learn More
Medchemexpress LLC PD153035 (hydrochloride) | 183322-45-4 | 99.5% | 10 MG
SDP
Catalog No. 5000233382
Encompass_Preferred
PD153035 Hydrochloride is a potent ATP-competitive epidermal growth factor receptor (EGFR) inhibitor used in biochemical and cellular research to block EGFR signaling with very high affinity.
  • Potent EGFR inhibitor; Ki ≈ 6 pM, IC50 ≈ 25 pM.
  • High reported purity of 99.5%.
  • Molecular weight 396.67 g·mol⁻¹; chemical formula C16H15BrClN3O2.
  • Available as a 10 mg solid and as 10 mM solutions in DMSO.
  • Suitable for biochemical assays and cell signaling studies.
Catalog No. 50-002-33382 Supplier Medchemexpress LLC Supplier No. HY1201310MG
PD153035 Hydrochloride is a potent ATP-competitive epidermal growth factor receptor (EGFR) inhibitor used in biochemical and cellular research to block EGFR signaling with very high affinity.
  • Potent EGFR inhibitor; Ki ≈ 6 pM, IC50 ≈ 25 pM.
  • High reported purity of 99.5%.
  • Molecular weight 396.67 g·mol⁻¹; chemical formula C16H15BrClN3O2.
  • Available as a 10 mg solid and as 10 mM solutions in DMSO.
  • Suitable for biochemical assays and cell signaling studies.