Learn More
Medchemexpress LLC PD153035 hydrochloride | 183322-45-4 | 99.5% | 100 MG
SDP
Catalog No. 5000231747
Encompass_Preferred
PD153035 hydrochloride is the hydrochloride salt of PD153035, a potent and selective epidermal growth factor receptor (EGFR) inhibitor (Ki 6 pM; IC50 25 pM) supplied as an off-white to light yellow solid for laboratory research.
  • Potent EGFR inhibitor with Ki 6 pM and IC50 25 pM.
  • High purity suitable for research applications.
  • Off-white to light yellow solid form.
  • Suitable for biochemical and cell-based assays.
  • Recommended storage: sealed, away from moisture at 4°C; in solvent: -80°C (six months), -20°C (one month).
Catalog No. 50-002-31747 Supplier Medchemexpress LLC Supplier No. HY12013100MG
PD153035 hydrochloride is the hydrochloride salt of PD153035, a potent and selective epidermal growth factor receptor (EGFR) inhibitor (Ki 6 pM; IC50 25 pM) supplied as an off-white to light yellow solid for laboratory research.
  • Potent EGFR inhibitor with Ki 6 pM and IC50 25 pM.
  • High purity suitable for research applications.
  • Off-white to light yellow solid form.
  • Suitable for biochemical and cell-based assays.
  • Recommended storage: sealed, away from moisture at 4°C; in solvent: -80°C (six months), -20°C (one month).