Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PD153035 hydrochloride | 183322-45-4 | 99.5% | 50 MG
SDP

Catalog No. 5000233494
Encompass_Preferred

PD153035 hydrochloride is the hydrochloride salt of a potent epidermal growth factor receptor (EGFR) inhibitor used in biochemical and cellular research. It is supplied as a high-purity, well-characterized small-molecule reagent with defined solubility and storage recommendations.

  • Potent EGFR inhibitor with reported Ki = 6 pM and IC50 = 25 pM.
  • High purity: 99.48%.
  • Molecular formula C16H15BrClN3O2; molecular weight 396.67 g/mol.
  • Soluble in DMSO (4 mg/mL); insoluble in water (< 0.1 mg/mL).
  • Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month).
  • Supplied as a 50 mg package for research use.

Catalog No. 50-002-33494 Supplier Medchemexpress LLC Supplier No. HY1201350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PD153035 hydrochloride is the hydrochloride salt of a potent epidermal growth factor receptor (EGFR) inhibitor used in biochemical and cellular research. It is supplied as a high-purity, well-characterized small-molecule reagent with defined solubility and storage recommendations.

  • Potent EGFR inhibitor with reported Ki = 6 pM and IC50 = 25 pM.
  • High purity: 99.48%.
  • Molecular formula C16H15BrClN3O2; molecular weight 396.67 g/mol.
  • Soluble in DMSO (4 mg/mL); insoluble in water (< 0.1 mg/mL).
  • Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month).
  • Supplied as a 50 mg package for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.