Learn More
TARGETMOL CHEMICALS INC PD158780 10MG
SDP
Catalog No. 502576686
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD 158780 reversibly inhibits auto and transphosphorylation of all four members of the ErbB receptor superfamily EGFR ErbB2 ErbB3 and ErbB4 (IC50s 8uM 49 nM 52 nM and 52 nM in cell assay). purity: 99%
Catalog No. 50-257-6686 Supplier TARGETMOL CHEMICALS INC Supplier No. T541010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD 158780 reversibly inhibits auto and transphosphorylation of all four members of the ErbB receptor superfamily EGFR ErbB2 ErbB3 and ErbB4 (IC50s 8uM 49 nM 52 nM and 52 nM in cell assay). purity: 99%