Learn More
Medchemexpress LLC HY-10321 10mg , PD173074 CAS:219580-11-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1032110MG
Encompass_Preferred
HY-10321 10mg PD173074 CAS: 219580-11-7 PD173074 is a potent FGFR1 inhibitor with an IC50 of 25 nM and also inhibits VEGFR2 with an IC50 of 100-200 nM, showing 1000-fold selectivity for FGFR1 over PDGFR and c-Src. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9219