Learn More
Selleck Chemical LLC PD173074 5mg 219580-11-7
SDP
Supplier:  Selleck Chemical LLC S12645MG
Encompass_Preferred
PD173074 is a potent FGFR1 inhibitor with IC50 of ~25 nM and also inhibits VEGFR2 with IC50 of 100-200 nM in cell-free assays, ~1000-fold selective for FGFR1 than PDGFR and c-Src. PD173074 reduces proliferation and promotes apoptosis in gastric cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-691-1