Learn More
Apexbio Technology LLC PD184352 (CI-1040), 100mg. Cas: 212631-79-3 MFCD: MFCD02683961. Selective MEK inhibitor
SDP
Supplier:  Apexbio Technology LLC A1792100
Encompass_Preferred
PD184352 (also known as CI-1040), a benzhydroxamate derivative, is a potent and highly selective MEK1/2, two members of the family of MAPKKs, inhibitor that inhibits purified MEK1 with IC50 of 17 nM in a non-ATP and non-ERK1/2 competitive manner [1] Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4708