Learn More
Apexbio Technology LLC PD184352 (CI-1040) 212631-79-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063507
Encompass_Preferred
PD184352 (CI-1040 CAS 212631-79-3) is a benzhydroxamate derivative that selectively inhibits the MEK1/2 kinases members of the MAP kinase kinase (MAPKK) family It interacts with a hydrophobic pocket adjacent to the Mg-ATP binding region of MEK1/2 triggering conformational shifts that favor the inactive non-phosphorylated state of these enzymes Acting via an ATP- and ERK1/2-independent inhibitory mechanism PD184352 robustly suppresses purified MEK1 enzyme activity (IC50 17 nM) Preclinical research demonstrates efficacy in reducing tumor growth notably in murine xenograft models of colon carcinoma underscoring its utility as a research tool in oncology
Catalog No. 50-000-63507 Supplier Apexbio Technology LLC Supplier No. A17925.1
PD184352 (CI-1040 CAS 212631-79-3) is a benzhydroxamate derivative that selectively inhibits the MEK1/2 kinases members of the MAP kinase kinase (MAPKK) family It interacts with a hydrophobic pocket adjacent to the Mg-ATP binding region of MEK1/2 triggering conformational shifts that favor the inactive non-phosphorylated state of these enzymes Acting via an ATP- and ERK1/2-independent inhibitory mechanism PD184352 robustly suppresses purified MEK1 enzyme activity (IC50 17 nM) Preclinical research demonstrates efficacy in reducing tumor growth notably in murine xenograft models of colon carcinoma underscoring its utility as a research tool in oncology