Learn More
ACROBiosystems Recombinant Protein;Cynomolgus PD-1 / PDCD1 Protein, Fc Tag;HEK293;100UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Catalog No. 501659104
Encompass_Preferred
Recombinant Protein;Cynomolgus PD-1 / PDCD1 Protein, Fc Tag;100UG;Cynomolgus PD-1, Fc Tag (PD1-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. As a result of different glycosylation, the protein migrates as 55-66 kDa under reducing (R) condition, and 110-140 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human PD-L1, Mouse Fc Tag (Cat. No. PD1-H52A3) at 5 μg/mL (100 μL/well) can bind Cynomolgus PD-1, Fc Tag (Cat. No. PD1-C5254) with a linear range of 3-33 ng/mL (QC tested).
Catalog No. 50-165-9104 Supplier ACROBiosystems Supplier No. PD1C5254100UG
Recombinant Protein;Cynomolgus PD-1 / PDCD1 Protein, Fc Tag;100UG;Cynomolgus PD-1, Fc Tag (PD1-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. As a result of different glycosylation, the protein migrates as 55-66 kDa under reducing (R) condition, and 110-140 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human PD-L1, Mouse Fc Tag (Cat. No. PD1-H52A3) at 5 μg/mL (100 μL/well) can bind Cynomolgus PD-1, Fc Tag (Cat. No. PD1-C5254) with a linear range of 3-33 ng/mL (QC tested).