Learn More
ACROBiosystems Recombinant Protein;Rat PD-1 / PDCD1 Protein, Fc Tag;HEK293;1MG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Catalog No. 501658636
Encompass_Preferred
Recombinant Protein;Rat PD-1 / PDCD1 Protein, Fc Tag;1MG;Rat PD-1, Fc Tag (PD1-R5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_001100397.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8636 Supplier ACROBiosystems Supplier No. PD1R52531MG
Recombinant Protein;Rat PD-1 / PDCD1 Protein, Fc Tag;1MG;Rat PD-1, Fc Tag (PD1-R5253) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_001100397.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE).