Learn More
ACROBiosystems Recombinant Protein;Mouse PD-1 / PDCD1 Protein, Fc Tag;HEK293;1MG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Supplier:  ACROBiosystems PD1M52591MG
Encompass_Preferred
Recombinant Protein;Mouse PD-1 / PDCD1 Protein, Fc Tag;1MG;Mouse PD-1, Fc Tag (PD1-M5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_032824).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse PD-L1, His Tag (Cat. No. PD1-M5220) at 10 μg/mL (100 μl/well) can bind Mouse PD-1, Fc Tag (Cat. No. PD1-M5259) with a linear range of 0.16-5 μg/mL (QC tested).
Catalog No. 50-165-8167