Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human PD-1 / PDCD1, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Supplier:  ACROBiosystems PD1H82F125UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human PD-1 / PDCD1, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human PD-1, Fc Tag (PD1-H82F1) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 44.4 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Fc Tag (Cat. No. PD1-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Fc Tag (Cat. No. PD1-H82F1) with a linear range of 20-320 ng/mL (QC tested).
Catalog No. 50-165-8251