Learn More
ACROBiosystems Recombinant Protein;Rabbit PD-1 / PDCD1 Protein, Fc Tag;HEK293;100UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Catalog No. 501658658
Encompass_Preferred
Recombinant Protein;Rabbit PD-1 / PDCD1 Protein, Fc Tag;100UG;Rabbit PD-1, Fc Tag (PD1-RB5251) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # G1SUF0).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse PD-L1, His Tag (Cat. No. PD1-M5220) at 10 μg/mL (100 μL/well) can bind Rabbit PD-1, Fc Tag (Cat. No. PD1-RB5251) with a linear range of 3-50 ng/mL (QC tested).
Catalog No. 50-165-8658 Supplier ACROBiosystems Supplier No. PD1RB5251100UG
Recombinant Protein;Rabbit PD-1 / PDCD1 Protein, Fc Tag;100UG;Rabbit PD-1, Fc Tag (PD1-RB5251) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # G1SUF0).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse PD-L1, His Tag (Cat. No. PD1-M5220) at 10 μg/mL (100 μL/well) can bind Rabbit PD-1, Fc Tag (Cat. No. PD1-RB5251) with a linear range of 3-50 ng/mL (QC tested).