Learn More
ACROBiosystems Recombinant Protein;Rabbit PD-1 / PDCD1 Protein;HEK293;100UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Supplier:  ACROBiosystems PD1R52H0100UG
Encompass_Preferred
Recombinant Protein;Rabbit PD-1 / PDCD1 Protein;100UG;Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # G1SUF0).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.95 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8659