Learn More
ACROBiosystems Recombinant Protein;Human PD-1 / PDCD1 Protein, Llama IgG2b Fc Tag, low endotoxin;HEK293;100UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Supplier:  ACROBiosystems PD1H5259100UG
Encompass_Preferred
Recombinant Protein;Human PD-1 / PDCD1 Protein, Llama IgG2b Fc Tag, low endotoxin;100UG;Human PD-1, Llama IgG2b Fc Tag, low endotoxin (PD1-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 43.8 kDa. The protein migrates as 55-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Mouse IgG1 Fc Tag (Cat. No. PD1-H52A3) at 0.5 μg/mL (100 μL/well) can bind Human PD-1, Llama IgG2b Fc Tag, low endotoxin (Cat. No. PD1-H5259) with a linear range of 20-313 ng/mL (QC tested).
Catalog No. 50-165-7948