Learn More
Cayman Chemical PD1PDL1 InhIbItr 3trIfl 500ug
SDP
Catalog No. 502913291
Encompass_Preferred
A PD-1/PD-L1 interaction inhibitor (IC50 = 9 nM in a TR-FRET assay)
Catalog No. 50-291-3291 Supplier Cayman Chemical Supplier No. 36719500 UG
Add to Cart
Add to Cart
A PD-1/PD-L1 interaction inhibitor (IC50 = 9 nM in a TR-FRET assay)