Learn More
Marketlab PD240CH-Z4 MRI Hand-Held Ferromagnetic Detector
Description
- 4.7 in. W x 15.9 in. L x 1.6 in. H.
- Dual-sensing capabilities for separately screening either ferrous metallic items or any type of metallic item.
- Nonmagnetic construction is ideal for operation in Zone IV.
- Three modes of operation: high-performance ferrous only, detection around the head and body, or full metal detection around the entire body.
- Large search area for quick, accurate screening.
- Dual-tone and dual-color display for high-precision pinpointing.
- Mitigates the risk of an RF burn occurring in the scanner due to ferrous or nonferrous metals.
- Audible and visual-magnitude alert settings.
- Visual signaling combined with the audio provide feedback on the location and position of the object under alarm.
- Battery life: >100 hours for all metals; >50 hours for body or head.
- MR conditional to 3 Tesla.
Specifications
Specifications
| Languages | English |