Learn More
Lovibond™ PD250 Dispenser Set 1, VARIO DPD Free Chlorine
Catalog No. 09050182 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
09-050-182 10 mL
1 options
Catalog No. 09-050-182 Supplier Lovibond™ Supplier No. 194901
Add to Cart
Add to Cart
The dispenser set is designed for easy and controlled dosage of DPD powder reagents. One click gives the exact amount of reagent required for a 10 mL sample. The PD 250 is the perfect alternative to Powder Packs for those carrying out a number of tests, saving time and reducing waste.
  • The reagent is supplied in sealed glass vials, sufficient for up to 250 tests
  • Vials can be changed quickly and easily
  • Once the vial has been opened, the contents should be used within 6 months
Specifications
pH Range 0 to 14
Volume (Metric) 10 mL
Type Powder Dispenser