Learn More
Description
- Faster operation than powder packs and tablets
- Accurate reagent dosing
- Use within 6 months of opening
- For use with the PD250 dispenser
Specifications
Specifications
| pH Range | 0 to 14 |
| Volume (Metric) | 10 mL |
| Type | Total Chlorine DPD |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.