Learn More
Apexbio Technology LLC PD318088 391210-00-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061862
Encompass_Preferred
PD318088 (CAS 391210-00-7) is an allosteric non-ATP competitive inhibitor of MEK1/2 kinase activity Structurally related to PD184352 it interacts adjacent to the ATP-binding site of MEK1 in proximity to residues Ile216 Phe209 Arg189 and Asp190 stabilizing a ternary complex involving MEK1/2 and MgATP This binding moderately increases the dissociation constant (Kd) for the MEK dimerization state from 75 nM to 140 nM PD318088 is mainly utilized as a molecular tool in protein kinase signaling research particularly in pathways involving MEK1/2
Catalog No. 50-000-61862 Supplier Apexbio Technology LLC Supplier No. A57075.1
PD318088 (CAS 391210-00-7) is an allosteric non-ATP competitive inhibitor of MEK1/2 kinase activity Structurally related to PD184352 it interacts adjacent to the ATP-binding site of MEK1 in proximity to residues Ile216 Phe209 Arg189 and Asp190 stabilizing a ternary complex involving MEK1/2 and MgATP This binding moderately increases the dissociation constant (Kd) for the MEK dimerization state from 75 nM to 140 nM PD318088 is mainly utilized as a molecular tool in protein kinase signaling research particularly in pathways involving MEK1/2