Learn More
Apexbio Technology LLC PD318088, 5mg. Cas: 391210-00-7 MFCD: MFCD18384973. Allosteric MEK1/2 inhibitor, non-ATP competitive
SDP
Supplier:  Apexbio Technology LLC A57075
Encompass_Preferred
PD318088 is an allosteric and non-ATP competitive MEK1/2 inhibitor.PD318088 is an analog of PD184352, which means it might have important anti-proliferative activity against cancer cells, although no functional study of PD318088 is currently available. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4961