Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PD98059

Catalog No. PHZ1164
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHZ1164 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHZ1164 Supplier Invitrogen™ Supplier No. PHZ1164
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

PD98059 is a potent and selective cell permeable inhibitor of MAP kinase kinase (MEK). It is provided in a Lyophilized format.

PD98059 selectively blocks the activation of MEK, thereby inhibiting the phosphorylation and the activation of MAP kinase.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5604, 5605, 5606, 5607, 5608, 5609, 6416
Molecular Weight (g/mol) 267.3 Da
Purification Method HPLC
Quantity 5 mg
Sterility Non-sterile
Content And Storage Store in freezer (-5 to -30°C).
Inhibitor Properties Chemical
Inhibitors Kinase Inhibitor, Kinase Inhibitor (MAP Kinase Inhibitor)
Shipping Condition Dry Ice
Specific Inhibit or Target MEK (MAP kinase kinase)
Form Solid
Research Category Signal Transduction
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.