Learn More
Apexbio Technology LLC PD98059 167869-21-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063820
Encompass_Preferred
PD98059 (CAS 167869-21-8) is a selective reversible inhibitor of MAP kinase kinase (MEK/MAPK/ERK kinase) It specifically targets MEK1 affecting both basal and partially activated forms with a reported IC50 of approximately 10 M Treatment with PD98059 inhibits ERK1/2 phosphorylation in a dose-dependent manner resulting in downregulation of cyclin E/Cdk2 and cyclin D1/Cdk4 complexes In cell-based studies such as human leukemia U937 cell models treatment leads to cell growth arrest at the G1 phase and induction of apoptosis PD98059 is widely employed in research investigating signaling pathways involving ERK activation cell cycle regulation and apoptosis
Catalog No. 50-000-63820 Supplier Apexbio Technology LLC Supplier No. A16635.1
PD98059 (CAS 167869-21-8) is a selective reversible inhibitor of MAP kinase kinase (MEK/MAPK/ERK kinase) It specifically targets MEK1 affecting both basal and partially activated forms with a reported IC50 of approximately 10 M Treatment with PD98059 inhibits ERK1/2 phosphorylation in a dose-dependent manner resulting in downregulation of cyclin E/Cdk2 and cyclin D1/Cdk4 complexes In cell-based studies such as human leukemia U937 cell models treatment leads to cell growth arrest at the G1 phase and induction of apoptosis PD98059 is widely employed in research investigating signaling pathways involving ERK activation cell cycle regulation and apoptosis