Learn More
Apexbio Technology LLC PD98059, 50mg. Cas: 167869-21-8 MFCD: MFCD00671789. MEK inhibitor,selective and reversible
SDP
Supplier:  Apexbio Technology LLC A166350
Encompass_Preferred
PD98059 is a selective and reversible inhibitor of MAPK-activating enzyme, MAPK/ERK kinase (MEK) that inhibits either basal MEK (GST-MEK1) or a partially activated MEK produced by mutation of serine to glutamate at 218 and 222 residues (GST-MEK-2E) with IC50 values of 10uM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4702