Learn More
Parker Hannifan PDA Agar Prefilled Plates, Designed for Compressed Air Testing, Compatible with CAMTU, 5 Plates
SDP
Catalog No. NC1253355
Encompass
The Parker Hannifin PDA Agar Prefilled Plates are specialized tools for monitoring microbial contamination in compressed air systems. Designed for use with the Parker Balston CAMTU, these plates facilitate the quick cultivation of yeast and mold, ensuring hygiene in sensitive environments like food processing. Their ready-to-use format and compatibility with Good Manufacturing Practices make them a reliable solution for air purity analysis.
Catalog No. NC1253355 Supplier Parker Hannifan Supplier No. C010134
The Parker Hannifin PDA Agar Prefilled Plates are specialized tools for monitoring microbial contamination in compressed air systems. Designed for use with the Parker Balston CAMTU, these plates facilitate the quick cultivation of yeast and mold, ensuring hygiene in sensitive environments like food processing. Their ready-to-use format and compatibility with Good Manufacturing Practices make them a reliable solution for air purity analysis.