Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, PDA/TUV Detectors, Intelligent Chip, Annual
Catalog No. NC1242024 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This performance maintenance kit for PDA with Intelligent Chip Technology contains all the quality parts necessary to keep the instrument operating at peak performance on an annual basis. It is used in UPLC PDA Detectors, eLambda PDA Detector, and UPLC TUV Detectors utilizing Intelligent Chip Technology. The kit includes the PerformancePLUS Long Life Lamp, which is warranted for a lifetime of 2000 hours or 1 year, whichever comes first.
  • Contains all quality parts necessary to keep the instrument operating at peak performance.
  • Designed for an annual replacement schedule.
  • Compatible with UPLC PDA Detectors, eLambda PDA Detector, and UPLC TUV Detectors.
  • Utilizes intelligent chip technology.
  • Includes a PerformancePLUS Long Life Lamp.
  • Lamp warranted for a lifetime of 2000 hours or 1 year.
Catalog No. NC1242024 Supplier Waters Corp Supplier No. 201000273
This performance maintenance kit for PDA with Intelligent Chip Technology contains all the quality parts necessary to keep the instrument operating at peak performance on an annual basis. It is used in UPLC PDA Detectors, eLambda PDA Detector, and UPLC TUV Detectors utilizing Intelligent Chip Technology. The kit includes the PerformancePLUS Long Life Lamp, which is warranted for a lifetime of 2000 hours or 1 year, whichever comes first.
  • Contains all quality parts necessary to keep the instrument operating at peak performance.
  • Designed for an annual replacement schedule.
  • Compatible with UPLC PDA Detectors, eLambda PDA Detector, and UPLC TUV Detectors.
  • Utilizes intelligent chip technology.
  • Includes a PerformancePLUS Long Life Lamp.
  • Lamp warranted for a lifetime of 2000 hours or 1 year.