Learn More
MOLTOX Molecular Toxicology Inc Potato Dextrose Agar w/Tartaric Acid, 20mL/plate, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948664
Encompass_Preferred
This media is used for the cultivation and enumeration of yeasts and molds. Potato Dextrose Agar is also used for the maintenance of stock cultures of certain dermatophytes and for the differentiation of varieties of dermatophytes via pigment production. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-664 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S32.141
This media is used for the cultivation and enumeration of yeasts and molds. Potato Dextrose Agar is also used for the maintenance of stock cultures of certain dermatophytes and for the differentiation of varieties of dermatophytes via pigment production. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.