Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDAM (1-Pyrenyldiazomethane)

Catalog No. P1405
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1405 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1405 Supplier Invitrogen™ Supplier No. P1405
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

PDAM (1-Pyrenyldiazomethane)

The fluorescent diazomethyl derivative PDAM (1-pyrenylyldiazomethane) has been commonly used to derivatize biomolecules. Diazoalkane react with the addition of catalysis and may be used for direct carboxylic acid modification of proteins and synthetic polymers. Fluorescent diazoalkanes also react with phosphates. PDAM is recommended as an alternative to ADAM because it has much better chemical stability. Moreover, the detection limit for PDAM for conjugates is reported to be about 20-30 femtomoles, which is five times better than reported for ADAM conjugates.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type PDAM
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Carboxylic Acid
Shipping Condition Wet Ice
Reactive Moiety Diazoalkane
Label or Dye Pyrene
Label Type Classic Dyes
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.