Learn More
Strem, An Ascensus Company CAS# 887919-35-9. Dichlorobis{[4-(N,N-dimethylamino)phenyl]di-t-butylphosphino}palladium(II), min. 98% PdAmphos. MFCD09265123. 5G
SDP
Catalog No. 501959545
Encompass
CAS# 887919-35-9. 5G. Dichlorobis{[4-(N,N-dimethylamino)phenyl]di-t-butylphosphino}palladium(II), min. 98% PdAmphos. MFCD09265123. Molecular Formula: C32H56Cl2N2P2Pd. Molecular Weight: 708.07. Color/Form: yellow pwdr. Strem#: 46-0825 www.strem.com/catalog/v3/46-0825/
Catalog No. 50-195-9545 Supplier Strem, An Ascensus Company Supplier No. 4608255G
CAS# 887919-35-9. 5G. Dichlorobis{[4-(N,N-dimethylamino)phenyl]di-t-butylphosphino}palladium(II), min. 98% PdAmphos. MFCD09265123. Molecular Formula: C32H56Cl2N2P2Pd. Molecular Weight: 708.07. Color/Form: yellow pwdr. Strem#: 46-0825 www.strem.com/catalog/v3/46-0825/