Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5133RBM4P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: PDCD1/1410R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG. Positive Control: TY cells or Tonsil. Cellular Staining: Cell Surface & Cytoplasmic. Buffer: 10mM PBS with or without 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human recombinant PDCD1 protein fragment. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55kDa.Chromosome: 2q37.3. Gene Symbol: PDCD1. Gene ID: 5133. Ensemble: ENSG00000188389. Uniprot: NP_016859782.
Catalog No. 50-188-5041